Learn More
Revvity Health Sciences Inc Revvity Cas9-gRNA Globin Depletion Enzyme, Human, HBA1, HBA2, HBB, HBD Targeting
Catalog No. NC3993771
Encompass_Preferred
This standalone, flexible CRISPR-Cas9 module is designed to remove globin-derived sequences from human blood RNA-seq libraries. It can be integrated into existing bulk or single-cell RNA-seq workflows at the double-stranded DNA stage, which reduces hemoglobin background and improves transcriptome coverage by directing reads toward informative transcripts.
  • Standalone, flexible CRISPR-Cas9 module
  • Removes globin-derived sequences from human blood RNA-seq libraries
  • Integrates into existing bulk or single-cell RNA-seq workflows at the double-stranded DNA stage
  • Reduces hemoglobin background for improved transcriptome coverage
  • Supplied as a pre-complexed Cas9-guide RNA mixture
  • Contains 30 guide RNAs targeting HBA1, HBA2, HBB, and HBD loci
  • Highly cost-competitive and widely compatible
  • Enables efficient globin depletion without requiring a specific library prep kit
Catalog No. NC3993771 Supplier Revvity Health Sciences Inc Supplier No. NOVA802500
Add to Cart
Add to Cart
This standalone, flexible CRISPR-Cas9 module is designed to remove globin-derived sequences from human blood RNA-seq libraries. It can be integrated into existing bulk or single-cell RNA-seq workflows at the double-stranded DNA stage, which reduces hemoglobin background and improves transcriptome coverage by directing reads toward informative transcripts.
  • Standalone, flexible CRISPR-Cas9 module
  • Removes globin-derived sequences from human blood RNA-seq libraries
  • Integrates into existing bulk or single-cell RNA-seq workflows at the double-stranded DNA stage
  • Reduces hemoglobin background for improved transcriptome coverage
  • Supplied as a pre-complexed Cas9-guide RNA mixture
  • Contains 30 guide RNAs targeting HBA1, HBA2, HBB, and HBD loci
  • Highly cost-competitive and widely compatible
  • Enables efficient globin depletion without requiring a specific library prep kit