Learn More
Revvity Health Sciences Inc Revvity Cas9-gRNA Mito Depletion Enzyme, Human, Pre-complexed, 657 Guide RNAs
Catalog No. NC3993772
Encompass_Preferred
The NEXTFLEX™ Cas9-gRNA Mito Depletion Enzyme (Human) is a flexible CRISPR-Cas9 module for removing mitochondrial derived sequences from human NGS libraries. It integrates into existing ATAC-seq, DNA-seq, bulk RNA-seq, or single-cell workflows at the double-stranded DNA stage, reducing mitochondrial background and focusing reads on nuclear genes and regulatory elements.
  • Supplied as a pre-complexed Cas9–guide RNA mixture.
  • Contains 657 guide RNAs targeting 17 mtDNA-encoded genes and selected nuclear mitochondrial pseudogenes.
  • One unit is defined as the amount required to deplete 1–10 ng of target double-stranded DNA library of at least 400 bp.
  • Delivers efficient mitochondrial depletion across diverse human library types.
  • Designed for broad compatibility without locking the user into a particular library preparation kit or sequencing platform.
Catalog No. NC3993772 Supplier Revvity Health Sciences Inc Supplier No. NOVA803100
The NEXTFLEX™ Cas9-gRNA Mito Depletion Enzyme (Human) is a flexible CRISPR-Cas9 module for removing mitochondrial derived sequences from human NGS libraries. It integrates into existing ATAC-seq, DNA-seq, bulk RNA-seq, or single-cell workflows at the double-stranded DNA stage, reducing mitochondrial background and focusing reads on nuclear genes and regulatory elements.
  • Supplied as a pre-complexed Cas9–guide RNA mixture.
  • Contains 657 guide RNAs targeting 17 mtDNA-encoded genes and selected nuclear mitochondrial pseudogenes.
  • One unit is defined as the amount required to deplete 1–10 ng of target double-stranded DNA library of at least 400 bp.
  • Delivers efficient mitochondrial depletion across diverse human library types.
  • Designed for broad compatibility without locking the user into a particular library preparation kit or sequencing platform.