Learn More
Revvity Health Sciences Inc Revvity Health Sciences Cas9-gRNA Mito Depletion Enzyme, Human, CRISPR-Cas9 Module
Catalog No. NC3998894
Encompass_Preferred
The NEXTFLEX Cas9-gRNA Mito Depletion Enzyme (Human) is a flexible CRISPR-Cas9 module that removes mitochondrial DNA from human NGS libraries. It integrates at the double-stranded DNA stage of various workflows, directing more reads to nuclear genes and regulatory elements by reducing mitochondrial background.
  • Removes mitochondrial DNA sequences from human NGS libraries.
  • Integrates directly into existing ATAC-seq, DNA-seq, bulk RNA-seq, or single-cell workflows.
  • Directs more sequencing reads to nuclear genes, regulatory elements, and other loci of interest.
  • Supplied as a pre-complexed Cas9–guide RNA mixture.
  • Contains 657 guide RNAs targeting 17 mtDNA-encoded genes and nuclear pseudogenes.
  • Compatible with diverse human library types and sequencing platforms.
Catalog No. NC3998894 Supplier Revvity Health Sciences Inc Supplier No. NOVA803500
The NEXTFLEX Cas9-gRNA Mito Depletion Enzyme (Human) is a flexible CRISPR-Cas9 module that removes mitochondrial DNA from human NGS libraries. It integrates at the double-stranded DNA stage of various workflows, directing more reads to nuclear genes and regulatory elements by reducing mitochondrial background.
  • Removes mitochondrial DNA sequences from human NGS libraries.
  • Integrates directly into existing ATAC-seq, DNA-seq, bulk RNA-seq, or single-cell workflows.
  • Directs more sequencing reads to nuclear genes, regulatory elements, and other loci of interest.
  • Supplied as a pre-complexed Cas9–guide RNA mixture.
  • Contains 657 guide RNAs targeting 17 mtDNA-encoded genes and nuclear pseudogenes.
  • Compatible with diverse human library types and sequencing platforms.