Learn More
Revvity Health Sciences Inc Revvity Health Sciences Inc. HTRF Detection Kit, Human and Mouse TGFβ1, 500 Assay Points
Catalog No. NC3983274
Encompass
The HTRF human and mouse TGFβ1 kit is designed for the quantification of human or mouse TGFβ1 release in cell supernatant.
  • Designed for the quantification of human or mouse TGFβ1 release in cell supernatant.
  • Includes reagents for acid activation step, which is required to detect TGFβ1 in samples and cell culture supernatants.
  • Antibodies labeled with donor and acceptor are pre-mixed and added in a single dispensing step.
  • Assay can be run up to a 1536 well-format by resizing addition volumes proportionally.
  • No cross-reactivity with human TGFβ2 and TGFβ3.
Catalog No. NC3983274 Supplier Revvity Health Sciences Inc Supplier No. 62HTGFBPEG
The HTRF human and mouse TGFβ1 kit is designed for the quantification of human or mouse TGFβ1 release in cell supernatant.
  • Designed for the quantification of human or mouse TGFβ1 release in cell supernatant.
  • Includes reagents for acid activation step, which is required to detect TGFβ1 in samples and cell culture supernatants.
  • Antibodies labeled with donor and acceptor are pre-mixed and added in a single dispensing step.
  • Assay can be run up to a 1536 well-format by resizing addition volumes proportionally.
  • No cross-reactivity with human TGFβ2 and TGFβ3.