Learn More
ABclonal Technology REXO2 Rabbit pAb
SDP
Catalog No. 5000108701
Encompass_Preferred
This gene encodes a 3-to-5 exonuclease specific for small (primarily 5 nucleotides or less in length) single-stranded RNA and DNA oligomers. This protein may have a role in DNA repair, replication, and recombination, and in RNA processing and degradation. It may also be involved in resistance of human cells to UV-C-induced cell death through its role in the DNA repair process.
Catalog No. 50-001-08701 Supplier ABclonal Technology Supplier No. A15808500UL
This gene encodes a 3-to-5 exonuclease specific for small (primarily 5 nucleotides or less in length) single-stranded RNA and DNA oligomers. This protein may have a role in DNA repair, replication, and recombination, and in RNA processing and degradation. It may also be involved in resistance of human cells to UV-C-induced cell death through its role in the DNA repair process.