Learn More
Aviva Systems Biology Corp REXO4 Antibody - N-terminal region (ARP39415_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39415_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against REXO4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human REXO4. Alternate names: REX4, XPMC2, XPMC2HApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-57024