Learn More
Gyros Inc REXXIP A-MAX
SDP
Catalog No. NC9920016
Encompass
2x standard formulation sample diluent. Buffer choice for non-primate unless analytes are positively charged or hydrophobic or if heterophilic antibodies are present in the sample.
Catalog No. NC9920016 Supplier Gyros Inc Supplier No. P0004821
2x standard formulation sample diluent. Buffer choice for non-primate unless analytes are positively charged or hydrophobic or if heterophilic antibodies are present in the sample.