Learn More
Gyros Inc REXXIP A
SDP
Catalog No. NC9920015
Encompass
1x standard formulation sample diluent. Buffer choice for non-primate unless analytes are positively charged or hydrophobic or if heterophilic antibodies are present in the sample.
Catalog No. NC9920015 Supplier Gyros Inc Supplier No. P0004820
1x standard formulation sample diluent. Buffer choice for non-primate unless analytes are positively charged or hydrophobic or if heterophilic antibodies are present in the sample.