Learn More
Gyros Inc REXXIP AN 25ML 1XSTD FORMULA.
SDP
Catalog No. NC9058587
Encompass
1x standard formulation sample diluent; increased ionic strength for positively charged analytes.
Catalog No. NC9058587 Supplier Gyros Inc Supplier No. P0004994
1x standard formulation sample diluent; increased ionic strength for positively charged analytes.