Learn More
Gyros Inc REXXIP AN-MAX BUFFER, 25 mL, 2x standard formulation, increased ionic strength, immunoassay buffer, enhances binding, minimizes non-specific binding, compatible with Gyrolab CD
SDP
Catalog No. NC0850006
Encompass
REXXIP AN-MAX BUFFER, 25 mL, 2x standard formulation, increased ionic strength, immunoassay buffer, enhances binding, minimizes non-specific binding, compatible with Gyrolab CD
Catalog No. NC0850006 Supplier Gyros Inc Supplier No. P0004995
REXXIP AN-MAX BUFFER, 25 mL, 2x standard formulation, increased ionic strength, immunoassay buffer, enhances binding, minimizes non-specific binding, compatible with Gyrolab CD