Learn More
Gyros Inc REXXIP CCS BUFFER 25ML
SDP
Catalog No. NC2046135
Encompass
Rexxip CCS25 mL, 1 x standard formulation 2. Option for cell culture sample with high protein content, e.g., bioprocess samples.
Catalog No. NC2046135 Supplier Gyros Inc Supplier No. P0004824
Rexxip CCS25 mL, 1 x standard formulation 2. Option for cell culture sample with high protein content, e.g., bioprocess samples.