Learn More
Gyros Inc REXXIP H
SDP
Catalog No. NC9920017
Encompass
1x standard formulation sample diluent. Agents to neutralizer heterophilic antibodies. For sampes containing heterophilic antibodies (primate) and wehere the assay format (Ab/Ab) could cause false positives.
Catalog No. NC9920017 Supplier Gyros Inc Supplier No. P0004822
1x standard formulation sample diluent. Agents to neutralizer heterophilic antibodies. For sampes containing heterophilic antibodies (primate) and wehere the assay format (Ab/Ab) could cause false positives.