Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gyros Inc Rexxip HN-MAX buffer, 25 mL, 2x standard formulation, for positively-charged analytes, neutralizes heterophilic antibodies, increases ionic strength
SDP

Catalog No. NC1459082
Encompass

The Gyros Rexxip HN-max is a specialized buffer designed for optimizing immunoassay performance within certain systems. This buffer is particularly effective for positively-charged analytes, offering properties that enhance assay accuracy and reliability. Key components of the formulation help neutralize unwanted antibodies and increase ionic strength, which together aid in preserving sample integrity and improving data quality in research settings.

  • Volume: 25 mL
  • Formulation: 2x standard formulation
  • Application: Positively-charged analytes
  • Special feature: Neutralizes heterophilic antibodies
  • Benefits: Preserves sample integrity
  • Benefits: Enhances analyte-reagent binding
  • Benefits: Minimizes non-specific binding
  • Benefits: Improves data quality
  • Benefits: Reduces assay development time
  • Quality: Batch-to-batch consistency
  • Regulatory status: Research use only

Catalog No. NC1459082 Supplier Gyros Inc Supplier No. P0004997
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gyros Rexxip HN-max is a specialized buffer designed for optimizing immunoassay performance within certain systems. This buffer is particularly effective for positively-charged analytes, offering properties that enhance assay accuracy and reliability. Key components of the formulation help neutralize unwanted antibodies and increase ionic strength, which together aid in preserving sample integrity and improving data quality in research settings.

  • Volume: 25 mL
  • Formulation: 2x standard formulation
  • Application: Positively-charged analytes
  • Special feature: Neutralizes heterophilic antibodies
  • Benefits: Preserves sample integrity
  • Benefits: Enhances analyte-reagent binding
  • Benefits: Minimizes non-specific binding
  • Benefits: Improves data quality
  • Benefits: Reduces assay development time
  • Quality: Batch-to-batch consistency
  • Regulatory status: Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.