Learn More
Gyros Inc REXXIP HX BUFFER
SDP
Catalog No. NC0273352
Encompass
1x Standard formulation sample diluent; agents to neutralize heterophilic antibodies. Suitable for human GLP-1 with potential for use with other negatively charged analytes.
Catalog No. NC0273352 Supplier Gyros Inc Supplier No. P0020033
1x Standard formulation sample diluent; agents to neutralize heterophilic antibodies. Suitable for human GLP-1 with potential for use with other negatively charged analytes.