Learn More
Med Vet International Reynold’s Equine Tooth Extractor, Upper, 15in, J0043RU, Each
Catalog No. 5000584578
Encompass_Preferred
This is the recommended instrument for retained deciduous teeth (caps) in horses. Also works for small premolars and incisors.
Catalog No. 50-005-84578 Supplier Med Vet International Supplier No. JOR43RU
This is the recommended instrument for retained deciduous teeth (caps) in horses. Also works for small premolars and incisors.