Learn More
Stoelting™ Reynolds Hemostatic Forceps
SDP
Catalog No. 10000477 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Full-Curved
Catalog No. Model
10-000-477 Full-Curved
1 options
Catalog No. 10-000-477 Supplier Stoelting™ Supplier No. 5211021P
Add to Cart
Add to Cart
Hemostatic forceps are used in surgical procedures to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
Specifications
Size 5.5 in.
Model Full-Curved