Learn More
Medchemexpress LLC Rezafungin acetate 10mM 1mL | 1631754-41-0 | 10MM 1ML
SDP
Catalog No. 5000241980
Encompass_Preferred
Rezafungin acetate (Biafungin acetate) is a next-generation broad-spectrum and long-lasting echinocandin Rezafungin acetate shows potent antifungal activity against Candida spp Aspergillus spp and Pneumocystis spp [1 [2
Catalog No. 50-002-41980 Supplier Medchemexpress LLC Supplier No. HY108009A10MM 1ML
Rezafungin acetate (Biafungin acetate) is a next-generation broad-spectrum and long-lasting echinocandin Rezafungin acetate shows potent antifungal activity against Candida spp Aspergillus spp and Pneumocystis spp [1 [2