Learn More
TARGETMOL CHEMICALS INC REZATOMIDINE 5MG
SDP
Catalog No. 502580268
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rezatomidine (AGN203818) is a selective antagonist of (alpha)2-adrenergic receptor and can be used in studies about chronic pain including neuropathic pain. purity: 99%
Catalog No. 50-258-0268 Supplier TARGETMOL CHEMICALS INC Supplier No. T343085MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rezatomidine (AGN203818) is a selective antagonist of (alpha)2-adrenergic receptor and can be used in studies about chronic pain including neuropathic pain. purity: 99%