Learn More
Cayman Chemical Rezivertinib, Purity: ≥98%, CAS: 1835667-12-3, 10 mg
SDP
Catalog No. 502978503
Encompass_Preferred
An inhibitor of mutant EGFR; selectively inhibits the proliferation of mutant-EGFR-containing PC-9, HCC827, and H1975 cells (GI50s = 13.3, 6.8, and 22 nM, respectively) over wild-type EGFR-containing A431 cells (GI50 = >1,000 nM); increases survival in an H1975 brain orthotopic rat model
Catalog No. 50-297-8503 Supplier Cayman Chemical Supplier No. 4253710 MG
An inhibitor of mutant EGFR; selectively inhibits the proliferation of mutant-EGFR-containing PC-9, HCC827, and H1975 cells (GI50s = 13.3, 6.8, and 22 nM, respectively) over wild-type EGFR-containing A431 cells (GI50 = >1,000 nM); increases survival in an H1975 brain orthotopic rat model