Learn More
MSC Menda Hand Lotion, ESD, Unscented, 32 oz Bottle
Supplier: MSC 89526396
This hand lotion reduces contamination that can cause solderability problems and helps lower skin electrical resistance, providing a reliable path-to-ground when using wrist straps, ESD gloves, or ESD footwear. It also restores moisture and provides relief to dry hands, which can be caused by continued use of nitrile or latex gloves and frequent washing.
- No fragrance and no dyes, ideal for any user.
- Silicone-free, non-contaminating, and free of chemicals that may adversely affect solderability or cause corrosion of electronic components.
- Dissipative ESD formulation for electronic manufacturing environments, will not insulate skin like regular lotions.
- Non-acidic.
- Absorbs quickly, not tacky or greasy.
- Contains vitamins A, D, E and aloe vera.
- Packaged in a 32 oz bottle.