Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc rF 710 aHu CD45RA Anti Primary

Catalog No. 5000086672
Encompass_Preferred

Product Name: redFluor 710 Anti-Human CD45RA (HI100)
Product Class: Antibody
Type: Primary
Product Name: redFluor 710 Anti-Human CD45RA (HI100)
Cytek Catalog Number: 80-0458-T025
Marker: CD45RA
Target Species: Human
Clonality: Monoclonal
Clone: HI100
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: redFluor 710
Size: 25 Tests

Catalog No. 50-000-86672 Supplier Cytek Biosciences Inc Supplier No. 800458T025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: redFluor 710 Anti-Human CD45RA (HI100)
Product Class: Antibody
Type: Primary
Product Name: redFluor 710 Anti-Human CD45RA (HI100)
Cytek Catalog Number: 80-0458-T025
Marker: CD45RA
Target Species: Human
Clonality: Monoclonal
Clone: HI100
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: redFluor 710
Size: 25 Tests

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.