Learn More
MOLECULAR DEPOT LLC RF and HAMA Blockers - 30 mL
Catalog No. 5000013494
Encompass_Preferred
ELISA Sample Diluent with RF and HAMA Blockers is a unique ELISA sample diluent specially formulated to block interference from both rheumatoid factors (RF) human anti-mouse antibodies (HAMA) This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13494 Supplier MOLECULAR DEPOT LLC Supplier No. B2015220
ELISA Sample Diluent with RF and HAMA Blockers is a unique ELISA sample diluent specially formulated to block interference from both rheumatoid factors (RF) human anti-mouse antibodies (HAMA) This product has been used as a molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only