Learn More
Glass Expansion RF Coil Alignment Tool Agilent 5000 Series ICP-OES
SDP
Catalog No. 502703058
Encompass_Preferred
RF Coil Alignment Tool Agilent 5000 Series ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent 5000 Series ICP-OES, P/Ns 501907335 and NC2081830.To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3058 Supplier Glass Expansion Supplier No. 708031415
RF Coil Alignment Tool Agilent 5000 Series ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent 5000 Series ICP-OES, P/Ns 501907335 and NC2081830.To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001