Learn More
Glass Expansion RF Coil Gold for Agilent 7500. To ensure correct installation, the Installation Kit 70-900-7500T is required. This is not included and must be ordered separately. It is re-useable so only needs to be ordered once.
SDP
Catalog No. 501907344
Encompass_Preferred
RF Coil Gold for Agilent 7500 ICP-MS. Compatible with Agilent 7500 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Silver 501907345.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 7500/7700/7800/7900/88/8900 (501907346).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7344 Supplier Glass Expansion Supplier No. 709007500G
RF Coil Gold for Agilent 7500 ICP-MS. Compatible with Agilent 7500 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Silver 501907345.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 7500/7700/7800/7900/88/8900 (501907346).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001