Learn More
Glass Expansion RF Coil Installation Tool for Varian Series 2 Axial, Suitable for: Liberty Series 2 and Vista.
SDP
Catalog No. 501907311
Encompass_Preferred
RF Coil Installation Tool for Agilent (Varian) Liberty Series 2 and Vista Axial ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent (Varian) Liberty Series 2 and Vista Axial ICP-OES, P/Ns 501907308, 501907309 and 501907310. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7311 Supplier Glass Expansion Supplier No. 709001000T
RF Coil Installation Tool for Agilent (Varian) Liberty Series 2 and Vista Axial ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent (Varian) Liberty Series 2 and Vista Axial ICP-OES, P/Ns 501907308, 501907309 and 501907310. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001