Learn More
Glass Expansion RF Coil Installation Tool for Optima 2000/4000/5000/7000
SDP
Catalog No. 501907329
Encompass_Preferred
RF Coil Installation Tool for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES, P/Ns 501907327 and 501907328. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7329 Supplier Glass Expansion Supplier No. 709002003T
RF Coil Installation Tool for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for PerkinElmer Optima 2000/4000/5000/7000 DV Series ICP-OES, P/Ns 501907327 and 501907328. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001