Learn More
Glass Expansion RF Coil Installation Tool for 700-ES Series Axial
SDP
Catalog No. 501907322
Encompass_Preferred
RF Coil Installation Tool for Agilent (Varian) 700-ES Series Axial ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent (Varian) 700-ES Series Axial ICP-OES, P/Ns 501907319, 501907320 and 501907321. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7322 Supplier Glass Expansion Supplier No. 709001004T
RF Coil Installation Tool for Agilent (Varian) 700-ES Series Axial ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent (Varian) 700-ES Series Axial ICP-OES, P/Ns 501907319, 501907320 and 501907321. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001