Learn More
Glass Expansion RF Coil Silver for Agilent 7700/8800/8900, To ensure correct installation, the Installation Kit 70-900-7500T is required. This is not included and must be ordered separately. It is re-useable so only needs to be ordered once.
SDP
Catalog No. 501907349
Encompass_Preferred
RF Coil Silver for Agilent 7700/8800/8900 ICP-MS. Compatible with all Agilent 7700/8800/8900 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Copper 501907347, Gold 501907348.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 75/77/78/79/88/8900 (501907346).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7349 Supplier Glass Expansion Supplier No. 709007700S
RF Coil Silver for Agilent 7700/8800/8900 ICP-MS. Compatible with all Agilent 7700/8800/8900 ICP-MS. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch, in addition to the power amplifier or solid-state generator. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Copper 501907347, Gold 501907348.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit for Agilent 75/77/78/79/88/8900 (501907346).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001