Learn More
Glass Expansion RF Coil Silver for Varian Series 2 Radial
SDP
Catalog No. 501907313
Encompass_Preferred
RF Coil Silver for Agilent (Varian) Liberty Series 2 and Vista Radial ICP-OES. Compatible with Agilent (Varian) Liberty Series 2 and Vista Radial ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Gold 501907312 and PTFE Coated Silver 501907314.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (501907315).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7313 Supplier Glass Expansion Supplier No. 709001001S
RF Coil Silver for Agilent (Varian) Liberty Series 2 and Vista Radial ICP-OES. Compatible with Agilent (Varian) Liberty Series 2 and Vista Radial ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. For Gold 501907312 and PTFE Coated Silver 501907314.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (501907315).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001