Learn More
MOLECULAR DEPOT LLC RF Plasma 1000 IU/mL 1 mL
Catalog No. 5000013565
Encompass_Preferred
Rheumatoid Factor Human Plasma is defibrinated human plasma containing high levels of RF (over 1000 IU/mL) No preservatives have been added This product is available in volumes ranging from a few mL to several Liters All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units Bulk volumes at discounted bulk prices are also available For in vitro research use only
Catalog No. 50-000-13565 Supplier MOLECULAR DEPOT LLC Supplier No. S2010001B
Rheumatoid Factor Human Plasma is defibrinated human plasma containing high levels of RF (over 1000 IU/mL) No preservatives have been added This product is available in volumes ranging from a few mL to several Liters All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units Bulk volumes at discounted bulk prices are also available For in vitro research use only