Learn More
MOLECULAR DEPOT LLC RF Purified 20000-24999 - 1 mL
Catalog No. 5000013532
Encompass_Preferred
High Rheumatoid Factor Solution Purified consists of a concentrated solution of human Rheumatoid Factors (20000-24999 IU/mL) that have been purified through several chromatography steps and supplied in buffer Titer has been measured with the RF Latex method This product can be used to test RF interference in immunoassays For in vitro research use only
Catalog No. 50-000-13532 Supplier MOLECULAR DEPOT LLC Supplier No. B2010861PF
High Rheumatoid Factor Solution Purified consists of a concentrated solution of human Rheumatoid Factors (20000-24999 IU/mL) that have been purified through several chromatography steps and supplied in buffer Titer has been measured with the RF Latex method This product can be used to test RF interference in immunoassays For in vitro research use only