Learn More
MOLECULAR DEPOT LLC RF Purified 7500-9999 - 1 mL
Catalog No. 5000013515
Encompass_Preferred
High Rheumatoid Factor Solution Purified consists of a concentrated solution of human Rheumatoid Factors (7500-9999 IU/mL) that have been purified through several chromatography steps and supplied in buffer Titer has been measured with the RF Latex method This product can be used to test RF interference in immunoassays For in vitro research use only
Catalog No. 50-000-13515 Supplier MOLECULAR DEPOT LLC Supplier No. B2010861PC
High Rheumatoid Factor Solution Purified consists of a concentrated solution of human Rheumatoid Factors (7500-9999 IU/mL) that have been purified through several chromatography steps and supplied in buffer Titer has been measured with the RF Latex method This product can be used to test RF interference in immunoassays For in vitro research use only