Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teledyne ISCO, Inc RF RACK 16 X 100MM 2/PK

Catalog No. 50201480 Shop All Teledyne ISCO Inc. Products
Encompass

CombiFlash Rf racks, 16 x 100 mm, qty 2 (108 tubes/rack)

Catalog No. 50-201-480 Supplier Teledyne ISCO, Inc Supplier No. 605237061
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CombiFlash Rf racks, 16 x 100 mm, qty 2 (108 tubes/rack)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.