Learn More
MOLECULAR DEPOT LLC RF Removal Kit - 10 reactions
Catalog No. 5000013463
Encompass_Preferred
Rheumatoid Factor Removal Kit is a kit designed to deplete human serum or plasma from Rheumatoid Factors (RF) This kit uses a proprietary solution to specifically and easily capture RF Simply add serum or plasma to the provided solution vortex spin down and recover your RF-free sample For in vitro research use only Not for diagnostic or therapeutic Use
Catalog No. 50-000-13463 Supplier MOLECULAR DEPOT LLC Supplier No. K2010013
Rheumatoid Factor Removal Kit is a kit designed to deplete human serum or plasma from Rheumatoid Factors (RF) This kit uses a proprietary solution to specifically and easily capture RF Simply add serum or plasma to the provided solution vortex spin down and recover your RF-free sample For in vitro research use only Not for diagnostic or therapeutic Use