Learn More
MOLECULAR DEPOT LLC RF Serum 15000-19999 - 1 mL
Catalog No. 5000013421
Encompass_Preferred
High Rheumatoid Factor Positive Serum is human serum containing high levels (15000-19999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only
Catalog No. 50-000-13421 Supplier MOLECULAR DEPOT LLC Supplier No. B2010861E
High Rheumatoid Factor Positive Serum is human serum containing high levels (15000-19999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only