Learn More
MOLECULAR DEPOT LLC RF Serum 2500-4999 - 1 mL
Catalog No. 5000013386
Encompass_Preferred
High Rheumatoid Factor Positive Serum is human serum containing high levels (2500-4999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only
Catalog No. 50-000-13386 Supplier MOLECULAR DEPOT LLC Supplier No. B2010861A
High Rheumatoid Factor Positive Serum is human serum containing high levels (2500-4999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only