Learn More
MOLECULAR DEPOT LLC RF Serum 7500-9999 - 1 mL
Catalog No. 5000013410
Encompass_Preferred
High Rheumatoid Factor Positive Serum is human serum containing high levels (7500-9999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only
Catalog No. 50-000-13410 Supplier MOLECULAR DEPOT LLC Supplier No. B2010861C
High Rheumatoid Factor Positive Serum is human serum containing high levels (7500-9999 IU/mL) of Rheumatoid Factor antibodies (RF) Titer has been measured with the RF Latex method No preservatives have been added All units have tested negative for infectious agents at the single donor level Available as single donor units or as pooled units For in vitro research use only