Learn More
Sigma Aldrich Fine Chemicals Biosciences Rf3000 Pipette Controller1EA
Catalog No. 502905062
Encompass
Powerful pump for quick and accurate measurements.The Rf3000 is an ergonomic lightweight pipette controller made of UV resistant material with adjustable valves and microprocessor control for consistent reliability and accuracy.The powerful pump offers rapid yet sensitive aspiration and dispensing. filling and dispensing speed is easily set with the same hand. Repeatedly press the large mode button and the bright LEDs will indicate one of three modes High Low or Gravity Mode (Gravity Mode to contain pipettes and dispense liquid without the motor).Additional features such as hand-neutral design for right- and left-handed users concave padded triggers and thumb controls ensures many hours of stress-free pipetting. The Rf3000 accommodates glass and plastic pipettes ranging in volume from 1 to 100 mL. The 0.45 A¬m nozzle filter prevents contamination and the 1mL nozzle adapter helps keep small pipettes stable.Long-life single Li-Ion battery allows approximately 20 hours of continuous
Catalog No. 50-290-5062 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7676701EA
Powerful pump for quick and accurate measurements.The Rf3000 is an ergonomic lightweight pipette controller made of UV resistant material with adjustable valves and microprocessor control for consistent reliability and accuracy.The powerful pump offers rapid yet sensitive aspiration and dispensing. filling and dispensing speed is easily set with the same hand. Repeatedly press the large mode button and the bright LEDs will indicate one of three modes High Low or Gravity Mode (Gravity Mode to contain pipettes and dispense liquid without the motor).Additional features such as hand-neutral design for right- and left-handed users concave padded triggers and thumb controls ensures many hours of stress-free pipetting. The Rf3000 accommodates glass and plastic pipettes ranging in volume from 1 to 100 mL. The 0.45 A¬m nozzle filter prevents contamination and the 1mL nozzle adapter helps keep small pipettes stable.Long-life single Li-Ion battery allows approximately 20 hours of continuous