Learn More
Medchemexpress LLC Rf470 TFA | 2369899-34-1 | 624.63 | 25 MG
SDP
Catalog No. 5000384219
Encompass_Preferred
Rf470 TFA is a FMR-probe-D-lysine conjugate (Max Ex: 470 nM; Max Em: 640 nM) that can be covalently incorporated into peptidoglycan by bacteria. It exhibits extremely weak fluorescence in its free state, but its fluorescence significantly increases when it is catalytically incorporated into peptidoglycan by transpeptidases.
  • Can be used for real-time monitoring of peptidoglycan biosynthesis.
  • Can be used for detection of transpeptidase activity.
  • Can be used for screening of antibiotics through fluorescence changes.
Catalog No. 50-003-84219 Supplier Medchemexpress LLC Supplier No. 25MGHY171995
Rf470 TFA is a FMR-probe-D-lysine conjugate (Max Ex: 470 nM; Max Em: 640 nM) that can be covalently incorporated into peptidoglycan by bacteria. It exhibits extremely weak fluorescence in its free state, but its fluorescence significantly increases when it is catalytically incorporated into peptidoglycan by transpeptidases.
  • Can be used for real-time monitoring of peptidoglycan biosynthesis.
  • Can be used for detection of transpeptidase activity.
  • Can be used for screening of antibiotics through fluorescence changes.