Learn More
Medchemexpress LLC HY-107382 1mg Medchemexpress, RF9 CAS:876310-60-0 Purity:>98%
SDP
Catalog No. 501968396
Encompass_Preferred
Medchemexpress, HY-107382 1mg RF9 CAS:876310-60-0 RF9 is a potent and selective Neuropeptide FF receptor antagonist, with Kis of 58±5 and 75±9 nM for hNPFF1R and hNPFF2R, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8396 Supplier Medchemexpress LLC Supplier No. HY1073821MG
Medchemexpress, HY-107382 1mg RF9 CAS:876310-60-0 RF9 is a potent and selective Neuropeptide FF receptor antagonist, with Kis of 58±5 and 75±9 nM for hNPFF1R and hNPFF2R, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.