Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RF9 hydrochloride | 00-00-0 | 99.9% | 519.08 | C26H39ClN6O3 | 1 ML
SDP

Catalog No. 5000240649
Encompass_Preferred

RF9 hydrochloride is a potent and selective antagonist of Neuropeptide FF (NPFF) receptors, with reported Ki values of 58 nM for hNPFF1R and 75 nM for hNPFF2R. It is supplied for research use as high-purity material, available as a 10 mM solution in DMSO or as dry powder in multiple vial sizes.

  • Potent NPFF receptor antagonist (Ki = 58 nM for hNPFF1R; Ki = 75 nM for hNPFF2R).
  • Molecular formula C26H39ClN6O3.
  • Molecular weight 519.08.
  • Purity >99.9%.
  • Available as 10 mM solution in DMSO and as dry powders in multiple vial sizes.
  • Intended for in vitro pharmacology and receptor binding studies.

Catalog No. 50-002-40649 Supplier Medchemexpress LLC Supplier No. HY107382A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RF9 hydrochloride is a potent and selective antagonist of Neuropeptide FF (NPFF) receptors, with reported Ki values of 58 nM for hNPFF1R and 75 nM for hNPFF2R. It is supplied for research use as high-purity material, available as a 10 mM solution in DMSO or as dry powder in multiple vial sizes.

  • Potent NPFF receptor antagonist (Ki = 58 nM for hNPFF1R; Ki = 75 nM for hNPFF2R).
  • Molecular formula C26H39ClN6O3.
  • Molecular weight 519.08.
  • Purity >99.9%.
  • Available as 10 mM solution in DMSO and as dry powders in multiple vial sizes.
  • Intended for in vitro pharmacology and receptor binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.