Learn More
Medchemexpress LLC RF9 hydrochloride | 00-00-0 | 99.9% | 519.1 g/mol | C26H39ClN6O3 | 5 MG
SDP
Catalog No. 5000239884
Encompass_Preferred
RF9 hydrochloride is the hydrochloride salt of RF9, a potent and selective antagonist of Neuropeptide FF (NPFF) receptors hNPFF1R and hNPFF2R. It is supplied for research use in pharmacology studies and is accompanied by standard quality documentation for laboratory handling.
  • Potent and selective NPFF receptor antagonist with Ki values of approximately 58 and 75 nM.
  • High purity appropriate for research applications.
  • Supplied as a stable solid suitable for formulation and dosing in preclinical studies.
  • Available in research-scale quantities (e.g., milligram levels).
  • Documentation available: certificate of analysis and safety data sheet.
Catalog No. 50-002-39884 Supplier Medchemexpress LLC Supplier No. HY107382A5MG
RF9 hydrochloride is the hydrochloride salt of RF9, a potent and selective antagonist of Neuropeptide FF (NPFF) receptors hNPFF1R and hNPFF2R. It is supplied for research use in pharmacology studies and is accompanied by standard quality documentation for laboratory handling.
  • Potent and selective NPFF receptor antagonist with Ki values of approximately 58 and 75 nM.
  • High purity appropriate for research applications.
  • Supplied as a stable solid suitable for formulation and dosing in preclinical studies.
  • Available in research-scale quantities (e.g., milligram levels).
  • Documentation available: certificate of analysis and safety data sheet.