Learn More
Medchemexpress LLC RF9 hydrochloride 10mg | 10MG
SDP
Catalog No. 5000239814
Encompass_Preferred
RF9 hydrochloride is a potent and selective Neuropeptide FF receptor antagonist with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R respectively[1 [2
Catalog No. 50-002-39814 Supplier Medchemexpress LLC Supplier No. HY107382A10MG
RF9 hydrochloride is a potent and selective Neuropeptide FF receptor antagonist with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R respectively[1 [2