Learn More
SAI Infusion Technologies Rat Femoral Artery Catheter - 30" tapered PU with a 3.3cm 2.5F tip
SDP
Catalog No. NC1211525
Encompass
Femoral artery catheter for Rats. 30" Tapered Polyurethane, 3.3cm 2.5Fr tip, accepts 19G or 20G blunt needle, 3 suture beads to anchor the catheter in place. Sterile. Product is for laboratory animal research only.
Catalog No. NC1211525 Supplier SAI Infusion Technologies Supplier No. RFA06
Femoral artery catheter for Rats. 30" Tapered Polyurethane, 3.3cm 2.5Fr tip, accepts 19G or 20G blunt needle, 3 suture beads to anchor the catheter in place. Sterile. Product is for laboratory animal research only.