Learn More
Aviva Systems Biology Corp RFC1 Antibody - middle region (ARP56522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RFC1. Alternate names: A1, MGC51786, MHCBFB, PO-GA, RECC1, RFC, RFC140ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-172