Learn More
Aviva Systems Biology Corp RFC2 Antibody - middle region (ARP61587_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61587_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFC2. It was validated on Western Blot. . Alternate names: A1, MGC3665, RFC40ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-413