Learn More
Aviva Systems Biology Corp RFC5 Antibody - N-terminal region (ARP46172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFC5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RFC5. Alternate names: MGC1155, RFC36ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-59258