Learn More
Aviva Systems Biology Corp RFESD Antibody - middle region (ARP55631_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55631_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFESD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RFESD. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61560