Learn More
Aviva Systems Biology Corp Rffl Antibody - N-terminal region (ARP42985_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42985_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rffl. It was validated on Western Blot. . Alternate names: 1700051E09Rik, 4930516L10Rik, BG080975, Carp2, MGC144763ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-049