Learn More
Aviva Systems Biology Corp RFFL Antibody - middle region (ARP42986_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42986_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFFL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RFFL. Alternate names: RNF189, RNF34L, CARP2, FRING, CARP-2, RIFIFYLINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63046